Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard Labels Vinyl BIOHAZARD

Catalog No. 17108809 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
108 ft. x 2 in.
2.875 x 2 in.
3.5 x 5 in.
5 x 3.5 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-108-809 108 ft. x 2 in.
17-108-798 2.875 x 2 in.
17-108-801 3.5 x 5 in.
17-108-799 5 x 3.5 in.
17-108-804 10 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-108-809 Supplier Brady™ Supplier No. 88051A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.

  • Good resistance to most common chemicals, oils and water
  • Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
  • Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
  • Service temperature of -40°F to +122°F (-40°C to +50°C)

Specifications

Adhesive Type Acrylic
Color Black on Orange
Finish Gloss
Format Tape
Material Vinyl
Product Type Barricade Tape
Sterility Non-sterile
Languages English
Quantity 1 Roll of 36 yd.
Shape Rectangle
Dimensions (L x W) 108 ft. x 2 in.
Length (English) 108 ft.
Width (English) 2 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.