Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Biohazard Pipet Keeper Container

Catalog No. 14375268 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
7.62 x 7.62 x 39.37 cm (3 x 3 x 15.5 in.)
8.89 x 11.43 x 41.91 cm (3.5 x 4.5 x 16.5 in.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
14-375-268 7.62 x 7.62 x 39.37 cm (3 x 3 x 15.5 in.)
21-000-123 8.89 x 11.43 x 41.91 cm (3.5 x 4.5 x 16.5 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-375-268
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Secure and convenient biohazard collection

  • Pipet keeper provides a reliable solution for collecting lab contaminants while keeping workstations tidy and compliant
  • Sturdy paperboard construction with visible biohazard symbols
  • Self-locking top and re-closable flaps for secure containment
  • Easily disposable after autoclaving per regulations
  • Cost-effective and user-friendly design

Specifications

Capacity (English) 2300 cc
Color White, Orange, Blue
Dimensions (L x W x H) 7.62 x 7.62 x 39.37 cm (3 x 3 x 15.5 in.)
Quantity 50/Cs.
Autoclavable Autoclavable
Material Plastic-lined Paperboard
Product Type Pipet Keeper
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.