Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard Sign Vinyl BIOHAZARD

Catalog No. 17108797 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
3.5 x 12 in.
3.5 x 5 in.
5 x 3.5 in.
7 x 10 in.
10 x 14 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-108-797 3.5 x 12 in.
17-108-808 3.5 x 5 in.
17-108-810 5 x 3.5 in.
17-108-806 7 x 10 in.
17-108-807 10 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-108-797 Supplier Brady™ Supplier No. 17753SLSA
Add to Cart
Add to Cart

Identify the actual or potential presence of a biologically hazardous material using Biohazard Labels made from self-sticking vinyl (B-585) material.

  • Good resistance to most common chemicals, oils and water
  • Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labelling
  • Warning labels support compliance with DOT 49 CFR 172.400(b) Labelling Requirements for transportation
  • Service temperature of -40°F to +122°F (-40°C to +50°C)

Specifications

Material Vinyl
Mounting Type Self-adhesive
Shape Rectangular
Languages English
Dimensions (L x W) 3.5 x 12 in.
Product Type BioHazard Safety Label
Color Black on Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.