Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Biohazard Specimen Bag

Catalog No. 12888306 Shop All Fisherbrand Products
Change view
Click to view available options
Thickness (English):
1.80 mil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (English)
12-888-306 1.80 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-888-306

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Fisherbrand™ Biohazard Specimen Bag is a three wall biohazard specimen bag designed for safely collecting, storing, and transporting specimen containers

  • Preprinted with orange and black Biohazard logo and storage instructions
  • Contains 2 x 3 in. absorbent pad to reduce contact with up to 3 mL of specimen fluids

Specifications

Closure Type Zipper
Color Clear
Includes 2 x 3 in. Absorbent Pad
Material LDPE
Sterility Non-sterile
No. per Case 6000/Cs.
No. per Pack 1000 Pk.
Thickness (English) 1.80 mil
Length (English) 6 in.
Width (English) 4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.