Learn More
Alkali Scientific Biohazard Specimen Transport Bag, 6"×9", LDPE, Tamper Evident Adhesive Closure with Seperate Document Pouch, 100/Pk, 1000/Case, with Absorbent Pad, Bottom Tear Line
SDP
Catalog No. 5000663199
Encompass_Preferred
This 6" × 9" Biohazard Specimen Transport Bag is manufactured from 2 mil gauge polyethylene (PE) and designed for the secure handling and transport of clinical and laboratory specimens. The glue closure provides a liquid tight, tamper evident seal to help protect specimen integrity during transit. Users simply peel off the liner to expose the adhesive and press to close, creating a reliable seal with visible evidence of tampering. The dual pouch design includes a rear pouch for specimen containment and an attached front document compartment for labels and paperwork, helping keep documentation separate from specimens. Clearly printed with the universal biohazard symbol, these bags are suitable for hospitals, diagnostic laboratories, clinics, and drug testing applications where safe specimen transport and clear identification are essential.
Catalog No. 50-006-63199 Supplier Alkali Scientific Supplier No. SB106X9APTL
This 6" × 9" Biohazard Specimen Transport Bag is manufactured from 2 mil gauge polyethylene (PE) and designed for the secure handling and transport of clinical and laboratory specimens. The glue closure provides a liquid tight, tamper evident seal to help protect specimen integrity during transit. Users simply peel off the liner to expose the adhesive and press to close, creating a reliable seal with visible evidence of tampering. The dual pouch design includes a rear pouch for specimen containment and an attached front document compartment for labels and paperwork, helping keep documentation separate from specimens. Clearly printed with the universal biohazard symbol, these bags are suitable for hospitals, diagnostic laboratories, clinics, and drug testing applications where safe specimen transport and clear identification are essential.