Learn More
Alkali Scientific Biohazard Specimen Transport Bag, Easy Open™ Zip Seal with Score Line & Seperate Document Pouch, 100/Pk, 1,000/Case, 6"×9"
SDP
Catalog No. 5000663292
Encompass_Preferred
These Biohazard Specimen Transport Bags are manufactured from durable 2 mil gauge LDPE (polyethylene) and are designed for the safe transport and handling of clinical and laboratory specimens. Available in multiple s, these bags accommodate a variety of specimen containers and transport requirements. Each bag features a dual pouch configuration, with a secure ziplock closure pouch for specimens and a separate document pouch to keep requisition forms or paperwork isolated from the sample. The bags are clearly printed with the universal biohazard symbol for quick identification and comply with OSHA and NCCLS guidelines for specimen transport. Select variants include an integrated absorbent pad that helps contain potential leaks during transit, providing additional safety and reliability for laboratories, hospitals, and diagnostic facilities.
Catalog No. 50-006-63292 Supplier Alkali Scientific Supplier No. SB06X9EZ69
These Biohazard Specimen Transport Bags are manufactured from durable 2 mil gauge LDPE (polyethylene) and are designed for the safe transport and handling of clinical and laboratory specimens. Available in multiple s, these bags accommodate a variety of specimen containers and transport requirements. Each bag features a dual pouch configuration, with a secure ziplock closure pouch for specimens and a separate document pouch to keep requisition forms or paperwork isolated from the sample. The bags are clearly printed with the universal biohazard symbol for quick identification and comply with OSHA and NCCLS guidelines for specimen transport. Select variants include an integrated absorbent pad that helps contain potential leaks during transit, providing additional safety and reliability for laboratories, hospitals, and diagnostic facilities.