Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caresfield Biohazard Specimen Transport Bag
SDP

Catalog No. 21222075
Change view
Click to view available options
Dimensions (L x W):
9 x 6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
21-222-075 9 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-222-075 Supplier Caresfield Supplier No. 48609SB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specimen transport bags provide a safe and compliant method of transporting specimens.

  • Biohazard symbols signal material risks
  • Colors and printed STAT messages signal correct specimen identification
  • Check boxes instruct the correct way to store specimen during transport
  • Usage is indicated on bag
  • Zip-top for easy and secure closure, and an optional tear-off top for fast specimen access
  • A separate back pouch is included on the back for storage of documents
  • Moisture-resistant bags protect specimen from environmental effects

Specifications

Autoclavable Not Autoclavable
Closure Type Tear-away Zip-top
Color Clear
Disposable Single-use
Includes Back pouch pocket for paperwork
Material Plastic
Sterility Non-sterile
Thickness (English) 0.0394 in.
Dimensions (L x W) 9 x 6 in.
Length (English) 9 in.
Product Type Specimen Transport Bag
Capacity (English) 4.55 lb.
Capacity (Metric) 5.01 L
Length (Metric) 22.86 cm
Width (Metric) 15.24 cm
Thickness (Metric) 1 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.