Learn More
Alkali Scientific Biohazard Specimen Transport Bags (Envelopes) , LDPE, Seperate Document Pouch, Tamper Evident Adhesive Closure, 100/Pk, 1000/Case, 6"x9"
SDP
Catalog No. 5000663337
Encompass_Preferred
These LDPE biohazard specimen transport bags are designed for the safe handling and transport of clinical and laboratory specimens. Manufactured from durable low density polyethylene, each bag features a clearly printed biohazard symbol for quick identification and regulatory compliance. A tamper evident adhesive closure creates a secure, liquid tight seal that helps prevent leaks and contamination during transport. The dual pouch design includes a sealed specimen compartment and a separate rear pouch for labels, forms, and documentation, helping keep paperwork clean and organized. A tear line at the top allows for easy opening after delivery while maintaining specimen security throughout transit, making these bags suitable for hospitals, diagnostic laboratories, clinics, and testing facilities.
Catalog No. 50-006-63337 Supplier Alkali Scientific Supplier No. SB206X9
These LDPE biohazard specimen transport bags are designed for the safe handling and transport of clinical and laboratory specimens. Manufactured from durable low density polyethylene, each bag features a clearly printed biohazard symbol for quick identification and regulatory compliance. A tamper evident adhesive closure creates a secure, liquid tight seal that helps prevent leaks and contamination during transport. The dual pouch design includes a sealed specimen compartment and a separate rear pouch for labels, forms, and documentation, helping keep paperwork clean and organized. A tear line at the top allows for easy opening after delivery while maintaining specimen security throughout transit, making these bags suitable for hospitals, diagnostic laboratories, clinics, and testing facilities.