Learn More
Alkali Scientific Biohazard Specimen Transport Bags, LDPE, Adhesive Closure with Tear Line, Liquid Tight, Separate Document Pouch, 12"×15", 1,000/Case, with Absorbent Pad
SDP
Catalog No. 5000663201
Encompass_Preferred
These Biohazard Specimen Transport Bags are manufactured from durable LDPE (polyethylene) and are designed for the secure transport and handling of clinical and laboratory specimens and bodily fluids. Primarily used by hospitals, laboratories, and anti doping agencies, they support safe specimen handling and regulatory compliance. Available in multiple options, the bags accommodate different specimen volumes and transport needs. Each bag features a liquid tight adhesive closure that forms a tamper evident seal once closed. A tear line enables easy opening while clearly indicating if the bag has been previously accessed. A separate front document pouch keeps labels and requisitions isolated from the specimen compartment. The bags display the universal biohazard symbol and comply with OSHA and NCCLS guidelines, with select variants including an integrated absorbent pad for additional leak protection.
Catalog No. 50-006-63201 Supplier Alkali Scientific Supplier No. SB312X15AP
These Biohazard Specimen Transport Bags are manufactured from durable LDPE (polyethylene) and are designed for the secure transport and handling of clinical and laboratory specimens and bodily fluids. Primarily used by hospitals, laboratories, and anti doping agencies, they support safe specimen handling and regulatory compliance. Available in multiple options, the bags accommodate different specimen volumes and transport needs. Each bag features a liquid tight adhesive closure that forms a tamper evident seal once closed. A tear line enables easy opening while clearly indicating if the bag has been previously accessed. A separate front document pouch keeps labels and requisitions isolated from the specimen compartment. The bags display the universal biohazard symbol and comply with OSHA and NCCLS guidelines, with select variants including an integrated absorbent pad for additional leak protection.