Learn More
Marketlab, Inc. Biohazard TearZone™ Bags
Description
- Bright orange and black print distinguishes stat specimens from routine specimens
- Three-wall construction and back flap protects all contents
- TearZone feature for easy opening
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Ziplock |
| Color | Clear with Orange Block |
| Disposable | Single-use |
| Material | LDPE |
| Sterility | Non-sterile |
| Thickness (English) | 2.0 mil |
| Dimensions (L x W) | 6 x 9 in. |
| Length (English) | 9 in. |
| Width (English) | 6 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.