Learn More
Electron Microscopy Sciences Tough-Spots BioHazard Label, 3/8"
SDP
Catalog No. 5034524
Encompass_Preferred
Tough-tags make labeling and identifying samples quick and easy are temperature resistant from -196 degree C to 80 degree C. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Size: 3/8"
Catalog No. 50-345-24 Supplier Electron Microscopy Sciences Supplier No. 7759314
Tough-tags make labeling and identifying samples quick and easy are temperature resistant from -196 degree C to 80 degree C. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Size: 3/8"