Learn More
Andwin Scientific BIOHAZARD WASTE BOX FLOOR 6PK
SDP
Catalog No. 50727627
Encompass_Preferred
The Floor Biohazard Waste Box features colorful graphics that allow fast identification. This product can be utilized to dispose ampoules, broken glass, vials. The boxes have cardboard outer material with inner 2-mil polypropylene bag. These measure 12 x 12 x 20". It is sold in a pack of 6.
Catalog No. 50-727-627 Supplier Andwin Scientific Supplier No. 64416
The Floor Biohazard Waste Box features colorful graphics that allow fast identification. This product can be utilized to dispose ampoules, broken glass, vials. The boxes have cardboard outer material with inner 2-mil polypropylene bag. These measure 12 x 12 x 20". It is sold in a pack of 6.