Learn More
Electron Microscopy Sciences Biohazard Waste Box, Intermediate, 12" x 12" x 20", 7" Opening
SDP
Catalog No. 5024823
Encompass_Preferred
These boxes are used for the disposal and incineration of biohazardous waste. The outer box is made from corrugated cardboard; the inner one is a 2-mil polypropylene bag. When the box is full, the safety cap is pulled into place and the entire unit is ready for incineration.
Dimensions: 12" x 12" x 20"
Size: 7" Opening
Catalog No. 50-248-23 Supplier Electron Microscopy Sciences Supplier No. 64415
These boxes are used for the disposal and incineration of biohazardous waste. The outer box is made from corrugated cardboard; the inner one is a 2-mil polypropylene bag. When the box is full, the safety cap is pulled into place and the entire unit is ready for incineration.
Dimensions: 12" x 12" x 20"
Size: 7" Opening