Learn More
Justrite™ Biohazard Waste Can, Foot-Operated Self-Closing SoundGard™ Cover
GSA_VA
Catalog No. 19822426 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Silver
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-426 White
19-822-425 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-426 Supplier Justrite™ Supplier No. 05915
  • $277.00
    $179.65 / Each of 1
    Save $97.35
    35% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment.
The Justrite Biohazard Waste Can, Foot-Operated Self-Closing SoundGard™ Cover provides a safe, temporary solution for collecting biological waste. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.
Justrite’s Biohazard Waste Containers meet OSHA 29 CFR 1910.1030 for the segregation of infectious waste. Use them to store contaminated laundry as well as other regulated waste; however, not for the storage of sharps. Sturdy steel cans feature leakproof construction with foot-operated, self-closing lid. Simple, foot pedal operation opens the cover automatically to safely dispose of biohazard waste. Upon release of the pedal, the can cover closes quickly to reduce the risk of contamination. A biohazard bag is held securely around the circumference of the can top.
Biohazard bags sold separately (No. 05901)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Capacity (Metric) 22 L
Capacity (English) 6 gal.
Color White