Learn More
Justrite™ Biohazard Waste Can, Foot-Operated Self-Closing SoundGard™ Cover
Catalog No. p-7070899 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Silver
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-425 Silver
19-822-426 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-425 Supplier Justrite™ Supplier No. 05914
Only null left
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment.
The Justrite Biohazard Waste Can, Foot-Operated Self-Closing SoundGard™ Cover provides a safe, temporary solution for collecting biological waste. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.
Justrite’s Biohazard Waste Containers meet OSHA 29 CFR 1910.1030 for the segregation of infectious waste. Use them to store contaminated laundry as well as other regulated waste; however, not for the storage of sharps. Sturdy steel cans feature leakproof construction with foot-operated, self-closing lid. Simple, foot pedal operation opens the cover automatically to safely dispose of biohazard waste. Upon release of the pedal, the can cover closes quickly to reduce the risk of contamination. A biohazard bag is held securely around the circumference of the can top.
Biohazard bags sold separately (No. 05901)
Specifications
Capacity (Metric) 22 L
Capacity (English) 6 gal.
Color Silver