Learn More
Sigma Aldrich Fine Chemicals Biosciences Biohazard waste container
Catalog No. 502900968
Encompass
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials. Convenient one-step sterilization no need to remove bag. Port opening in cover allows one-handed disposal of materials limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust dent or puncture.
Catalog No. 50-290-0968 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. TMO63700004
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials. Convenient one-step sterilization no need to remove bag. Port opening in cover allows one-handed disposal of materials limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust dent or puncture.