Learn More
Sigma Aldrich Fine Chemicals Biosciences Biohazard waste container1EA
Catalog No. 502905864
Encompass
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials. Convenient one-step sterilization no need to remove bag. Port opening in cover allows one-handed disposal of materials limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust dent or puncture.
Catalog No. 50-290-5864 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3706731EA
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials. Convenient one-step sterilization no need to remove bag. Port opening in cover allows one-handed disposal of materials limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust dent or puncture.