Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Medical Biohazardous Waste Bags
SDP

Catalog No. 22045353 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
38 x 45 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-045-353 38 x 45 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-045-353 Supplier Medegen Medical Supplier No. CF2359
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biohazardous Waste Bags are printed with Biohazardous/Infectious Waste and Biohazard Symbol.

  • Print/Label: Biohazardous/Infectious Waste, Biohazard Symbol

Specifications

Certifications/Compliance Meet ASTM Dart Impact/Elmendorf Tear and DOT CFR 49 173-197 Requirements
Material LLDPE
No. per Case 125/Cs.
Thickness (English) 1.75 mil
Dimensions (L x W) 38 x 45 in.
Length (English) 38 in.
Width (English) 45 in.
Product Type Biohazardous Waste Bag
Capacity (English) 44 gal.
Capacity (Metric) 166.6 L
Length (Metric) 96.5 cm
Width (Metric) 114.3 cm
Thickness (Metric) 0.05 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.