Learn More
BioSpec Products BIOHOMOGENIZER, 2.5 CM PROBE
SDP
Catalog No. NC0449044
Encompass_Preferred
The BioHomogenizer is a rotor-stator type tissue homogenizer designed to rapidly homogenize, disperse, and emulsify biological samples in aqueous media. Probe included.
Catalog No. NC0449044 Supplier BioSpec Products Supplier No. 1285
The BioHomogenizer is a rotor-stator type tissue homogenizer designed to rapidly homogenize, disperse, and emulsify biological samples in aqueous media. Probe included.