Learn More
Perkin Elmer US LLC Biokinetics Accessory
Supplier:  Perkin Elmer US LLC L2250145
Encompass_Preferred
This accessory is compatible with the LS 45/50/50B/55 fluorescence spectrometers. It requires Accessory PCB Kit for use with LS 45.
Catalog No. 50-211-2751