Learn More
MSC Biolife WoundSeal Powder, Topical, Instant Bleeding Control, Sterile, 4 Applications
Catalog No. 5000677492
Encompass
WoundSeal Powder is a topical powder designed to stop minor external bleeding. It works by forming an instant, protective scab when applied with pressure, effectively stopping blood flow. This powder requires the presence of blood to be effective.
  • Stops minor external bleeding instantly
  • Forms a protective seal (scab) in 30 seconds
  • One application covers a wound area up to 3.9 cm² (0.6 in²) to a depth of 2 mm
  • Formulated with hydrophilic polymer and potassium ferrate
  • Contents are sterile
  • Provided as single-use applications
Catalog No. 50-006-77492 Supplier MSC Supplier No. 32925265
WoundSeal Powder is a topical powder designed to stop minor external bleeding. It works by forming an instant, protective scab when applied with pressure, effectively stopping blood flow. This powder requires the presence of blood to be effective.
  • Stops minor external bleeding instantly
  • Forms a protective seal (scab) in 30 seconds
  • One application covers a wound area up to 3.9 cm² (0.6 in²) to a depth of 2 mm
  • Formulated with hydrophilic polymer and potassium ferrate
  • Contents are sterile
  • Provided as single-use applications