Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioLite™ Cell Culture Treated Dishes

Catalog No. 12556003 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
12-556-003 150 mm
12-556-000 35 mm
12-556-001 60 mm
12-556-002 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-556-003 Supplier Thermo Scientific™ Supplier No. 130183
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Dishes that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.

Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated dishes suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.

• Cell culture treated
• Sterile per ISO 11137, SAL 10-6
• USP Class VI and non-cytotoxic
• Non-pyrogenic

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 35 mL (Well)
Culture Area 148 sq. cm
Material Polyethylene
Shape Round
Sterility Sterile
Surface Treatment Cell Culture
Type Cell Culture Dish
Volume (Metric) Working 35 mL
No. per Case 120
No. per Pack 10
Height (Metric) 20 mm
Height (Metric) Interior 15.4 mm
Diameter (Metric) 150 mm
Show More Show Less
What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.