Learn More
Percival Scientific Biological Incubator, I30LH1, 9.6 cu. ft., horizontal IncuWhite LED lighting, 3 tiers, pan-type humidifier and electronic RH sensor, 5-44° C ±0.5°, IntellusUltra controller
SDP
Catalog No. NC3793454
Encompass_Preferred
The I30LH1 incubator is frequently used for mosquitos and other insects, low light algae growth, or low cost seed germination. IncuWhite LEDs are dimmable between 5-100% output with up to 125 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. A pan-type humidifier and standard humidity sensor provides additive humidity.Three white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.
Catalog No. NC3793454 Supplier Percival Scientific Supplier No. I30LH1
Add to Cart
Add to Cart
Request a Quote
The I30LH1 incubator is frequently used for mosquitos and other insects, low light algae growth, or low cost seed germination. IncuWhite LEDs are dimmable between 5-100% output with up to 125 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. A pan-type humidifier and standard humidity sensor provides additive humidity.Three white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.