Learn More
Percival Scientific Biological Incubator, I41LLVLH3, 37.2 cu. ft., horizontal and vertical IncuWhite LED lights, 4 tiers, pan-type hum., dehum., elec. RH sensor, 4-44° C ±0.5°, IntellusUltra control
SDP
Catalog No. NC3793646
Encompass_Preferred
The I41LLVLH3 incubator is frequently used for mosquitos, other insects, low light algae growth, or low cost seed germination. IncuWhite LEDs are dimmable between 5-100% output with up to 220 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Pan-type humidifier, dehumidifier, and standard sensor maintain optimal humidity. Four white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.
Catalog No. NC3793646 Supplier Percival Scientific Supplier No. I41LLVLH3
Add to Cart
Add to Cart
Request a Quote
The I41LLVLH3 incubator is frequently used for mosquitos, other insects, low light algae growth, or low cost seed germination. IncuWhite LEDs are dimmable between 5-100% output with up to 220 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Pan-type humidifier, dehumidifier, and standard sensor maintain optimal humidity. Four white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.