Learn More
Percival Scientific Biological Incubator, I41VLH6, 37.2 cu. ft., vertical IncuWhite LED lighting, 5 tiers, pan-type humidifier, dehum. and advanced RH sensor, 4-44° C ±0.5°, IntellusUltra controller
SDP
Supplier:  Percival Scientific I41VLH6
Encompass_Preferred
The I41VLH6 incubator is frequently used for mosquitos, other insects, low light algae growth, or low cost seed germination. IncuWhite LEDs are dimmable between 5-100% output with up to 120 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Pan-type humidifier, dehumidifier, and adv. sensor tightly control humidity. Five white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.
Catalog No. NC3793084
Add to Cart
Request a Quote