Learn More
Fine Science Tools Dumont #5 Forceps, Straight, 0.05 x 0.02 mm, 11 cm
SDP
Catalog No. NC9626652
Encompass
Style: #5/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.02mm/ Alloy / Material: Titanium/ Length: 11cm
Catalog No. NC9626652 Supplier Fine Science Tools Supplier No. 1125240
Style: #5/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.02mm/ Alloy / Material: Titanium/ Length: 11cm