Learn More
Tisch Scientific Biologix 3ml Sterile Transfer Pipettes, 7.5ml Capacity - 2,000/case, 30-0138A1
SDP
Catalog No. NC1878646
Encompass
Transfer liquids without worrying about cross-contamination with Biologix sterile 3ml transfer pipettes.
  • Sterile, DNase/RNase free
  • 2,000/case, individually wrapped
  • Safe use and convenient disposal
  • Pipettes have molded graduations at 0.25ml up to 3ml (22 drops/ml)
Catalog No. NC1878646 Supplier Tisch Scientific Supplier No. 300138A1
Transfer liquids without worrying about cross-contamination with Biologix sterile 3ml transfer pipettes.
  • Sterile, DNase/RNase free
  • 2,000/case, individually wrapped
  • Safe use and convenient disposal
  • Pipettes have molded graduations at 0.25ml up to 3ml (22 drops/ml)