Learn More
BIOLOGIX USA INC Biologix Cardboard Freezer Box, 36-Well, 15 mL, -196°C to 121°C
Catalog No. NC3995549
Encompass_Preferred
These cardboard freezer boxes are constructed from durable hard cardboard with a white outer coating, designed for secure storage of laboratory samples. The 36-well configuration is specifically sized to accommodate 15 mL centrifuge tubes. They are resistant to chemicals and can withstand temperatures from -196°C to 121°C.
  • Durable hard cardboard construction with white outer coating.
  • Chemically resistant to alcohols and mild organic solvents.
  • Equipped with a removable cover for easy marking.
  • Working temperature range from -196°C to 121°C, suitable for liquid nitrogen freezing in vapor phase.
  • Includes convenient cardboard dividers for efficient organization.
  • Compatible with various freezer models for broad applicability.
  • Non-sterile.
Catalog No. NC3995549 Supplier BIOLOGIX USA INC Supplier No. 901536
These cardboard freezer boxes are constructed from durable hard cardboard with a white outer coating, designed for secure storage of laboratory samples. The 36-well configuration is specifically sized to accommodate 15 mL centrifuge tubes. They are resistant to chemicals and can withstand temperatures from -196°C to 121°C.
  • Durable hard cardboard construction with white outer coating.
  • Chemically resistant to alcohols and mild organic solvents.
  • Equipped with a removable cover for easy marking.
  • Working temperature range from -196°C to 121°C, suitable for liquid nitrogen freezing in vapor phase.
  • Includes convenient cardboard dividers for efficient organization.
  • Compatible with various freezer models for broad applicability.
  • Non-sterile.