Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioMag™ Micro Particles, Magnetic, Carboxy Functionalized , MilliporeSigma™ Supelco™

Catalog No. 111003182 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3182 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3182 Supplier MilliporeSigma Supelco Supplier No. 0023910MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Content And Storage 2°C to 8°C
Product Line BioMag
Quantity 10 mL
Synonym Magnetic micro particles, carboxy functionalized

BioMag is a registered trademark of Polysciences, Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.