Learn More
BANGS LABORATORIES INC BioMagPlus Concanavalin A Magnetic Particles, Affinity-Based Separation, High Purity & Yield, 10 mL
Catalog No. NC1781147
Encompass
BioMagPlus Concanavalin A are magnetic particles designed for efficient separation and purification of glycosylated biomolecules. Their Concanavalin A coating specifically binds to carbohydrate structures, improving purity and yield in proteomics and clinical diagnostics. With a nominal mean diameter of ~1.5 µm and broad compatibility for mammalian and bacterial systems, they streamline workflows by enhancing specificity and reducing processing times, differentiating them from traditional separation methods.
Catalog No. NC1781147 Supplier BANGS LABORATORIES INC Supplier No. BP53110ML
BioMagPlus Concanavalin A are magnetic particles designed for efficient separation and purification of glycosylated biomolecules. Their Concanavalin A coating specifically binds to carbohydrate structures, improving purity and yield in proteomics and clinical diagnostics. With a nominal mean diameter of ~1.5 µm and broad compatibility for mammalian and bacterial systems, they streamline workflows by enhancing specificity and reducing processing times, differentiating them from traditional separation methods.