Learn More
Bioventures Inc BIOMARKER HIGH 10000-3000BP
Catalog No. 50272365
Encompass
MAPMARKER, 70-400BP, ROX Labeled (70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400bp), 8fmoles/band/ul Concentration
Catalog No. 50-272-365 Supplier Bioventures Inc Supplier No. M2
MAPMARKER, 70-400BP, ROX Labeled (70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400bp), 8fmoles/band/ul Concentration