Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MedSchenker™ BIOMARQ™ Biopsy Container
SDP

Catalog No. 23000016
Change view
Click to view available options
No. per Case:
25 Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
23-000-016 25 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-000-016 Supplier MedSchenker™ Supplier No. MBC20F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An easier and safer way to fixate biopsies, this biopsy container uses formalin to minimize exposure. Simply twist cap to release reagent contents. Ideal for multiple types of specimen analysis with a container size that is larger than most.

  • Lid with encapsulated formalin to help minimize exposure
  • Electron beam sterilized and virtually shatterproof
  • Semi-closed system and screw cap lid for safer and simpler fixation of biopsies
  • 10 mL of reagent volume, holds 20 mL of volume total
  • Reagent includes 4% Formaldehyde and <2.5% Methanol

Specifications

Product Type Biopsy Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Cylindrical
Sterility Irradiated
Autoclavable Not Autoclavable
Certifications/Compliance IATA Packaging Instruction 650
Closure Material Polypropylene
Closure Type Screw Cap
Color Clear
Diameter (Metric) 37.3 mm
For Use With (Application) Analyzing Biopsy Samples
For Use With (Equipment) Tissue Biopsy Kit
Height (English) 1.53 in.
Height (Metric) 38.9 mm
Holds 20 mL
Includes Container, Lid
No. per Case 25 Cs.
No. per Pack 200 Pk.
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.