Learn More
Research Products International Corp BioMasher II Disposable Micro-Tube Homogenizer, Sterile, Individually Wrapped, 50 per Pack
SDP
Catalog No. 501367690
Encompass
BioMasher II provides a convenient and efficient method for homogenizing small samples directly in a micro-tube. Unique closed system consists of a specially designed 1.5ml clear polypropylene micro-tube with a polyacetal pestle. The pestle has a molded-in shaft guard for added protection from hazardous substances and helps prevent sample loss. Both tube and pestle feature abrasive surface areas that ensure maximum grinding efficiency. This disposable system is ideal for homogenizing animal tissue, plant tissue and various other biological samples.
Catalog No. 50-136-7690 Supplier Research Products International Corp Supplier No. 199623
BioMasher II provides a convenient and efficient method for homogenizing small samples directly in a micro-tube. Unique closed system consists of a specially designed 1.5ml clear polypropylene micro-tube with a polyacetal pestle. The pestle has a molded-in shaft guard for added protection from hazardous substances and helps prevent sample loss. Both tube and pestle feature abrasive surface areas that ensure maximum grinding efficiency. This disposable system is ideal for homogenizing animal tissue, plant tissue and various other biological samples.