Learn More
Thermo Scientific™ BioMate™ 160 UV-Vis Spectrophotometer PROMO
Description
The BioMate UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience. Software includes pre-programmed life science methods such as Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays and OD600.
A full line of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control.
Features:
- 7-inch high-resolution touchscreen user interface
- Pre-programmed life science methods
- Networking capable (Wi-Fi adaptor available)
- Room light resistance for 'open-lid' operation
- Sipper accessory
- Removable, washable sample compartment liner with magnetic placement and hold-down
- Accommodates cells up to 100 mm pathlength
- (Optional) 8-position cell changer with switchable turret
- (Optional) 4-position cell changer (long path cells)
- (Optional) Peltier-thermostatted single cell holder (20°C to 60°C)
- (Optional) Accessory for test-tubes up to 25 mm diameter and 150 mm height
Designed for:
- Advanced teaching labs
- Research and developement departments
- Applications requiring high throughput or temperature control
Specifications
Specifications
| Baseline Flatness | ±0.002 A |
| Compartments | Accommodates (optional): 8-position cell changer, 4-position cell changer (long path cells), Peltier thermostatted cell holder (20°C to 60°C), Sipper accessory, Fiber optic probe coupler |
| Connections | Single USB-A, Duplex USB-A, Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, Print via USB, Ethernet or Wi-Fi USB adaptor available |
| Detector Type | Dual Silicon Photodiodes |
| Display | 7 in. High Definition Color Touchscreen, Tiltable, 800 x 1280 pixels |
| Dimensions (L x W x H) | 35.5 x 38.5 x 19.5 cm |
| Spectral Bandwidth | 2 nm |
| Wavelength Range | 190 to 1100 nm |
| Drift | (At 500 nm after 1 hour warmup) <0.0005 A/hr. |
| Electrical Requirements | External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.