Learn More
Trajan Scientific Biomedal Cap Crimp 20mm White PTFE/Trans Blue Silicone, 100 Pack
Catalog No. NC0506925
Encompass_Preferred
Cap Crimp 20mm 3.0mm White PTFE/Trans Blue Silicone Bimetal Magnetic With 8mm hole100 Pack
Catalog No. NC0506925 Supplier Trajan Scientific Supplier No. XAR20031536
Cap Crimp 20mm 3.0mm White PTFE/Trans Blue Silicone Bimetal Magnetic With 8mm hole100 Pack