Learn More
PHCbi Biomedical Freezer
Catalog No. 502720059
Encompass_Preferred
PHCbi brand 17.8 cu.ft (504 L) biomedical ECO freezer utilizes environmentally friendly hydrocarbon refrigerants. This unit leverage innovative variable -speed inverter compressors to deliver efficient cooling and responsive recovery time after door openings. Temperature Range'-20℃ to -30℃
Catalog No. 50-272-0059 Supplier PHCbi Supplier No. MDFMU539HLPA
Add to Cart
Add to Cart
Request a Quote
PHCbi brand 17.8 cu.ft (504 L) biomedical ECO freezer utilizes environmentally friendly hydrocarbon refrigerants. This unit leverage innovative variable -speed inverter compressors to deliver efficient cooling and responsive recovery time after door openings. Temperature Range'-20℃ to -30℃