Learn More
Beckman Coulter Pipette Tips
Catalog No. NC2258306 Shop All Beckman Coulter Products
Encompass_Preferred
Beckman Coulter pipette tips are high-quality plastic consumables designed for use with Beckman Coulter pipettes. They come in a variety of sizes and configurations to accommodate different volume ranges and pipetting techniques. The tips feature a universal fit for use across Beckman's pipette lines. They are manufactured from medical-grade virgin polypropylene in a cleanroom environment. Key benefits include a tight seal for accuracy and precision, smooth tip interior for reduced sample retention, and color-coding for easy size identification. The tips are sterilized and RNase/DNase free. They are a reliable choice for researchers performing sensitive procedures like PCR.
This item is not returnable. View return policy
Catalog No. NC2258306 Supplier Beckman Coulter Supplier No. B85971
Beckman Coulter pipette tips are high-quality plastic consumables designed for use with Beckman Coulter pipettes. They come in a variety of sizes and configurations to accommodate different volume ranges and pipetting techniques. The tips feature a universal fit for use across Beckman's pipette lines. They are manufactured from medical-grade virgin polypropylene in a cleanroom environment. Key benefits include a tight seal for accuracy and precision, smooth tip interior for reduced sample retention, and color-coding for easy size identification. The tips are sterilized and RNase/DNase free. They are a reliable choice for researchers performing sensitive procedures like PCR.