Learn More
Beckman Coulter Biomek i3 Automated Workstation (D34958)
Catalog No. NC3920935
Encompass_Preferred
The Biomek i3 automated workstation configured with all five Flex positions delivers maximum versatility and throughput for complex workflows. Each Flex position supports integration of accessories and devices—such as shakers, heaters, magnetic separation, and tube racks—enabling seamless execution of multi-step processes like NGS library prep, PCR setup, and cell-based assays. With this fully equipped configuration, the Biomek i3 offers unmatched flexibility, optimized deck space, and streamlined automation to accelerate your research while maintaining precision and reproducibility.
Catalog No. NC3920935 Supplier Beckman Coulter Supplier No. D34958
The Biomek i3 automated workstation configured with all five Flex positions delivers maximum versatility and throughput for complex workflows. Each Flex position supports integration of accessories and devices—such as shakers, heaters, magnetic separation, and tube racks—enabling seamless execution of multi-step processes like NGS library prep, PCR setup, and cell-based assays. With this fully equipped configuration, the Biomek i3 offers unmatched flexibility, optimized deck space, and streamlined automation to accelerate your research while maintaining precision and reproducibility.