Learn More
Beckman Coulter Biomek i3 Automated Workstation (D34963)
Supplier:  Beckman Coulter D34963
Encompass_Preferred
The Biomek i3 automated workstation configured for genomics extractions combines precision liquid handling with integrated shaking, heating, and cooling devices to support demanding sample preparation workflows. This setup enables efficient lysis, binding, washing, and elution steps for DNA and RNA extraction, ensuring optimal temperature control and thorough mixing for high-quality yields. With flexible deck positions and intuitive software, the Biomek i3 streamlines complex protocols, reduces manual intervention, and delivers reproducible results for downstream applications like NGS and PCR.
Catalog No. NC3920943