Learn More
Beckman Coulter Biomek i3 Benchtop Liquid Handler, 8-Channel with Enclosure (D22660)
Supplier:  Beckman Coulter D22660
Encompass_Preferred
The Beckman Coulter Biomek i3 benchtop liquid handler (D22660) is an 8-channel, tip-based automated workstation designed for flexible, high-throughput pipetting workflows. It provides a compact benchtop platform with an enclosed work area for contamination control, an arm-mounted touchscreen, and preinstalled software for method development and routine automation.
  • 8-channel fixed pipetting pod covering a 1-1,000 μL volume range.
  • Deck capacity of 15 pipettable positions and 5 flex positions.
  • Independent tip loading and select-tip functionality.
  • Enclosed workspace with optional HEPA filtration and UV light.
  • Arm-mounted touchscreen with wireless keyboard and mouse.
  • Compatible with P20, P50, P250, and P1000 disposable tips.
  • Operates on 100-240 VAC power with standard automation controller and monitor requirements.
  • Compact benchtop footprint with door-open access for service and setup.
Catalog No. NC3920933