Learn More
Fluotics Biomek iSeries FX NX comp P30 non-sterile
Catalog No. NC2195916
Encompass_Preferred
Beckman Coulter Biomek iSeries FX NX compatible P30, non-sterile, working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85739
This item is not returnable. View return policy
Catalog No. NC2195916 Supplier Fluotics Supplier No. BFXI30.NS
Beckman Coulter Biomek iSeries FX NX compatible P30, non-sterile, working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85739