Learn More
Fluotics Biomek iSeries FX NX comp P30 filter/sterile Type A (1536 Plate/Tip Washer accessible)
Catalog No. NC2195913
Encompass_Preferred
Beckman Coulter Biomek iSeries FX NX compatible P30, filter/sterile, Type A (1536 Plate/Tip Washer accessible), working volume: 20 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85749
This item is not returnable. View return policy
Catalog No. NC2195913 Supplier Fluotics Supplier No. BFXI30.F/A
Beckman Coulter Biomek iSeries FX NX compatible P30, filter/sterile, Type A (1536 Plate/Tip Washer accessible), working volume: 20 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85749