Learn More
Fluotics Biomek iSeries FX NX comp P50 sterile Type A (1536 Plate/Tip Washer accessible)
Catalog No. NC2195932
Encompass_Preferred
Beckman Coulter Biomek iSeries FX NX compatible P50, sterile, Type A (1536 Plate/Tip Washer accessible), working volume: 50 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85756
This item is not returnable. View return policy
Catalog No. NC2195932 Supplier Fluotics Supplier No. BFXI50.ST/A
Beckman Coulter Biomek iSeries FX NX compatible P50, sterile, Type A (1536 Plate/Tip Washer accessible), working volume: 50 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85756