Learn More
Fluotics Biomek iSeries FX NX comp P50 filter/sterile Type A (1536 Plate/Tip Washer accessible)
Catalog No. NC2195930
Encompass_Preferred
Beckman Coulter Biomek iSeries FX NX compatible P50, filter/sterile, Type A (1536 Plate/Tip Washer accessible), working volume: 40 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85760
This item is not returnable. View return policy
Catalog No. NC2195930 Supplier Fluotics Supplier No. BFXI50.F/A
Beckman Coulter Biomek iSeries FX NX compatible P50, filter/sterile, Type A (1536 Plate/Tip Washer accessible), working volume: 40 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85760