Learn More
Beckman Coulter Biomek Span-8 P50 Conductive Tips, with Barrier
Catalog No. NC0324660
Encompass
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing
Catalog No. NC0324660 Supplier Beckman Coulter Supplier No. B01091
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing