Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ BioMesh Biopsy Bags
SDP

Catalog No. 22222170
Change view
Click to view available options
Dimensions (L x W):
30 x 45 mm
45 x 75 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-222-170 30 x 45 mm
22-222-171 45 x 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-222-170 Supplier StatLab™ Supplier No. BBSM1000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Synthetic mesh and small pore design ensures unrestricted flow of processing reagents around specimens.

  • Bags secure small biopsies during processing
  • Resistant to solvents and acids

Specifications

Product Type Biopsy Bag
Material Polyester
Color White
Length (Metric) 45 mm
Width (Metric) 30 mm
Dimensions (L x W) 30 x 45 mm
Size Small
Quantity 1000/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.