Learn More
StatLab BioMesh Cassettes
Description
- Lids separate, ideal for small, fragile specimens
- Excellent for use with Tissue Guard processing gel
- Keep small biopsies and fragile specimens secure with these fine mesh cassettes that eliminate the need for messy sponges, bags and paper
- Rounded BioMesh edges ensure no tissue fragments are lost in the corners
Specifications
Specifications
| Quantity | 250 Pk. |
| Color | White |