Learn More
Bellco Glass, Inc. Biometer Flask Complete, 250mL
SDP
Catalog No. 501945565
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Designed for the measurement of microbial co2 production over time. Ideal for bio-remediation studies.. MADE IN THE USA All components are autoclaveable. Complete 250mL, 1 each.
Catalog No. 50-194-5565 Supplier Bellco Glass, Inc. Supplier No. 255600250
Manufactured from type 1 class a Borosilicate glass. Designed for the measurement of microbial co2 production over time. Ideal for bio-remediation studies.. MADE IN THE USA All components are autoclaveable. Complete 250mL, 1 each.