Learn More
Labrepco Inc BIOMIXER 3D FIXED SPD 12"X12"
SDP
Catalog No. NC0923097
Encompass
The BioMixer Nutating Shaker provides a 3-dimensional "nutating" motion for thorough, foam-free mixing in blood and centrifuge tubes, as well as in a variety of flat vessels. Comes standard with autoclavable dimples mats while optional flat mats are also available for mixing other vessel types. This mixer features maintenance free, brushless motors capable of loads up to 2kg and are safe for use in cold rooms and incubators. Features: • Includes dimpled mat; perfect for mixing blood and centrifuge tubes • Tri-directional motion for thorough, uniform mixing • Stacking platforms available • Cold room and incubator safe Specifications: • Speed Range: Fixed, 24 rpm • Tilt Angle: Fixed, +/- 20° • Platform Size: 12 x 12 in. / 30 x 30 cm • Load Capacity: 4 lbs (2.2kg) • Operating Temp. Range: +4°C to +65°C • Dimensions (WxDxH): 12 x 12 x 8 in. or 30 x 30 x 20 cm • Weight: 4.4lbs (2kg) • Electrical: 115V, 50-60Hz, 50W
Catalog No. NC0923097 Supplier Labrepco Inc Supplier No. B3D1320
The BioMixer Nutating Shaker provides a 3-dimensional "nutating" motion for thorough, foam-free mixing in blood and centrifuge tubes, as well as in a variety of flat vessels. Comes standard with autoclavable dimples mats while optional flat mats are also available for mixing other vessel types. This mixer features maintenance free, brushless motors capable of loads up to 2kg and are safe for use in cold rooms and incubators. Features: • Includes dimpled mat; perfect for mixing blood and centrifuge tubes • Tri-directional motion for thorough, uniform mixing • Stacking platforms available • Cold room and incubator safe Specifications: • Speed Range: Fixed, 24 rpm • Tilt Angle: Fixed, +/- 20° • Platform Size: 12 x 12 in. / 30 x 30 cm • Load Capacity: 4 lbs (2.2kg) • Operating Temp. Range: +4°C to +65°C • Dimensions (WxDxH): 12 x 12 x 8 in. or 30 x 30 x 20 cm • Weight: 4.4lbs (2kg) • Electrical: 115V, 50-60Hz, 50W