Learn More
Polysciences, Inc. Disposable closed system micro tissue homogenizer. The abrasive surfaces on the pestle tip and inner tube area ensure efficient grinding of samples
SDP
Catalog No. NC0796134
Encompass
Disposable closed system micro tissue homogenizer. The abrasive surfaces on the pestle tip and inner tube area ensure efficient grinding of samples
Catalog No. NC0796134 Supplier Polysciences, Inc. Supplier No. 25533100
Disposable closed system micro tissue homogenizer. The abrasive surfaces on the pestle tip and inner tube area ensure efficient grinding of samples